Please login to your online account to display your discounted pricing

KH 7, Tocris™

Selective soluble adenylyl cyclase inhibitor

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material KH 7
Assay Percent Range >99%
Molecular Formula C17H15BrN4O2S
Formula Weight 419.3
Synonym (±)-2-(1H-benzimidazol-2-ylthio)propanoic acid 2-[(5-bromo-2-hydroxyphenyl)methylene]hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

383410

 
r & d systems
383410
10mg Each for $205.70

383450

 
r & d systems
383450
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 419.3
Quantity 10mg
Chemical Name or Material KH 7
Assay Percent Range >99%
Molecular Formula C17H15BrN4O2S
CAS 330676-02-3
Synonym (±)-2-(1H-benzimidazol-2-ylthio)propanoic acid 2-[(5-bromo-2-hydroxyphenyl)methylene]hydrazide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C