Keystone Safety Blue Polypropylene Lab Coats, Elastic Wrists
Cool and comfortable polypropylene lab coats are ideal for nonhazardous areas where a low particulate barrier is needed.
$121.98 - $132.47
Specifications
Product Type General Purpose Lab Coat
Gender Unisex
Material Polypropylene
Color Blue
Closure Type Snaps
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-061-445
View Documents Promotion Details
Keystone Safety
LCOBENWMED
Medium
Case of 30 Each for $121.98
19-061-450
View Documents Promotion Details
Keystone Safety
LCOBENW4XL
4X-Large
Case of 30 Each for $132.47
19-061-448
View Documents Promotion Details
Keystone Safety
LCOBENW2XL
2X-Large
Case of 30 Each for $121.98
19-061-444
View Documents Promotion Details
Keystone Safety
LCOBENWSM
Small
Case of 30 Each for $121.98
19-061-449
View Documents Promotion Details
Keystone Safety
LCOBENW3XL
3X-Large
Case of 30 Each for $123.88
19-061-447
View Documents Promotion Details
Keystone Safety
LCOBENWXL
X-Large
Case of 30 Each for $121.98
19-061-446
View Documents Promotion Details
Keystone Safety
LCOBENWLG
Large
Case of 30 Each for $121.98
Description
Description
  • Lab coats provide protection against dirt, grime, and non-hazardous dry particulates
  • Non-woven polypropylene material is durable and low linting
  • Ideal for use in food manufacturing, light industrial, and medical environments
  • Full collar, elastic wrists
  • 3-pocket style available
  • Packaged 30 per case
  • Meets ANSI 101-1996
Specifications
Specifications
General Purpose Lab Coat
Polypropylene
Snaps
Single-use
None
Yes
ANSI 101-1996
1.4 oz.
Not Autoclavable
Unisex
Blue
Elastic
Non-sterile
Single
LCO-BE-NW
Food, Manufacturing, Light Industrial, Medical
40 g
30/Pk.
SDS
missing translation for 'documents'
missing translation for 'documents'