Keystone Safety Polypropylene Arm Sleevess DFS Item
Lightweight, breathable fabric provides economical protection
$43.19 - $43.19
Specifications
Color White
View More Specs
Products 1
Catalog Number Mfr. No. Material Size Size (English) Size (Metric) Price Quantity  
Catalog Number Mfr. No. Material Size Size (English) Size (Metric) Price Quantity  
19-068-480
View Documents
Keystone Safety
AGNWILARGE
Polypropylene Large 45.7 x 22.8 cm
Case of 100 Each for $43.19
Description
Description
A high-quality, low-cost solution to protect workers against dry particulates, dirt and grime.
  • Made of non-woven, 100% spunbond polypropylene fabric
  • 100% latex free
  • White color
  • Flame resistant
  • 45.7 × 22.86cm (18 × 9 in.)
Specifications
Specifications
White
SDS