Please login to your online account to display your discounted pricing

Alfa Aesar™ Ketolide Resistance Peptide

For cellular and molecular biology applications

Specifications

Product Type Ketolide Resistance Peptide
Molecular Weight 698.87
Storage Requirements -30° to -10°C
Sequence Met-Arg-Phe-Phe-Val
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66633LB0

 
alfa aesar™
J66633LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66633MCR

 
alfa aesar™
J66633MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Ketolide Resistance Peptide
Molecular Weight 698.87
Storage Requirements -30° to -10°C
Sequence Met-Arg-Phe-Phe-Val
Format Solid
Common Name M-R-F-F-V
Regulatory Status RUO
Purification Method HPLC