Please login to your online account to display your discounted pricing

α-Ketoglutaric Acid Disodium Salt, Anhydrous, MP Biomedicals™

1,3-Lysolecithin is a class of phospholipids that are intermediates in the metabolism of lipids

$1,273.59 - $1,273.59

Chemical Identifiers

Chemical Name or Material α-Ketoglutaric Acid Disodium Salt, Anhydrous
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10005890

 
mp biomedicals
0210005890
500mg Each for $1,273.59
Description & Specifications

Specifications

Quantity 500mg
Chemical Name or Material α-Ketoglutaric Acid Disodium Salt, Anhydrous