Please login to your online account to display your discounted pricing

Alfa Aesar™ Ketoconazole, 2mg/mL in methanol, sterile-filtered

Chemical Identifiers

Chemical Name or Material Ketoconazole
CAS 65277-42-1
Name Note 2mg/mL in methanol, sterile-filtered
MDL Number MFCD00058579
UN Number UN1230
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ673528EQ

 
alfa aesar™
J673528EQ
5 x 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67352XF

 
alfa aesar™
J67352XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ketoconazole
Physical Form Liquid
MDL Number MFCD00058579
CAS 65277-42-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H311-H331-H360-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
May damage fertility or the unborn child.
Causes damage to organs.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
Name Note 2mg/mL in methanol, sterile-filtered
UN Number UN1230
Quantity 5 x 1mL
Storage Note 1 -30° to -10°C
RTECS Number TK7912300
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H331-H360-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
May damage fertility or the unborn child.
Causes damage to organs.

-30° to -10°C