Please login to your online account to display your discounted pricing

Ketanserin tartrate, Tocris

Selective 5-HT2A antagonist; Also antagonist at 5-HT1D

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material Ketanserin tartrate
Assay Percent Range >97%
Molecular Formula C22H22FN3O3·C4H6O6
Formula Weight 545.52
Synonym 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2, 4[1H, 3H]-quinazolinedione tartrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

0908250

 
r & d systems
0908250
50mg Each for $585.69

090850

 
r & d systems
090850
50mg Each for $139.08
Description & Specifications

Specifications

Chemical Name or Material Ketanserin tartrate
CAS 83846-83-7
Recommended Storage Store at Room Temperature
Assay Percent Range >97%
Molecular Formula C22H22FN3O3·C4H6O6
Formula Weight 545.52
Synonym 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2, 4[1H, 3H]-quinazolinedione tartrate
Quantity 50mg
Safety and Handling

Store at Room Temperature