missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ketanserin tartrate, 98+%

Selective 5-HT2A serotonin receptor antagonist

$587.55 - $587.55

Chemical Identifiers

CAS 83846-83-7
Molecular Formula C26H28FN3O9
Molecular Weight (g/mol) 545.52
MDL Number MFCD00084651
InChI Key KMTLTEVOQLMYRS-WUUYCOTASA-N
Synonym ketanserin tartrate
PubChem CID 91885469
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid;3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione
SMILES C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=O.C(C(C(=O)O)O)(C(=O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62798MD View Documents Alfa Aesar™
J62798MD
250mg Each for $587.55
 
Specifications

Chemical Identifiers

83846-83-7
545.52
KMTLTEVOQLMYRS-WUUYCOTASA-N
91885469
C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=O.C(C(C(=O)O)O)(C(=O)O)O
C26H28FN3O9
MFCD00084651
ketanserin tartrate
(2S,3S)-2,3-dihydroxybutanedioic acid;3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione

Specifications

White
83846-83-7
MFCD00084651
14,5296
Soluble in DMSO at 50mg/ml; Also soluble in 0.1N HCl (6mg/ml) or in ethanol (3.3mg/ml)
C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=O.C(C(C(=O)O)O)(C(=O)O)O
545.52
545.52
≥98%
250mg
C26H28FN3O9
UN2811
ketanserin tartrate
KMTLTEVOQLMYRS-WUUYCOTASA-N
(2S,3S)-2,3-dihydroxybutanedioic acid;3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione
91885469
Powder
Ketanserin tartrate
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : VA1411195

Recommended Storage : 2° to 8°C