Please login to your online account to display your discounted pricing

Kerosene Blank, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 1 gal.
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  KERBLKG

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. KER-BLK-G

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1 gal.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Chemical Name or Material Kerosene Blank
Packaging Triple-leached LDPE bottle
Health Hazard 1 Causes severe skin burns and eye damage
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Solvent or Matrix Kerosene
DOT Information Hazardous
Recommended Storage Ambient
Certificate of Analysis With Certificate of Analysis.

Specifications

Chemical Name or Material Kerosene Blank
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Accessories for Catalog Number: KERBLKG

Item Mfr. No. Item Description Price Per Unit Quantity  
296355104
Spilkleen/Cep  CEP LAB SPILL KIT SOLVENTS
spilkleen/cep
CSKLSK1SOL
Each for $341.99
Accessories

Causes severe skin burns and eye damage

Ambient