missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kerosene, Odorless, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 611030010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Kerosene | |
4.5 (Air = 1.0) | |
1 L | |
5mmHg at 38°C |
8008-20-6 | |
MFCD00135561 | |
15, 5342 |
Chemical Identifiers
8008-20-6 | |
MFCD00135561 |
Safety and Handling
DOTInformation : DOT Class 3
EINECSNumber : 232-366-4
RTECSNumber : OA5500000
TSCA : TSCA
RUO – Research Use Only