Please login to your online account to display your discounted pricing

Alfa Aesar™ Kerosene, low odor


Quantity 500mL

Manufacturer: alfa aesar™  L14479AP

 View more versions of this product

Catalog No. AAL14479AP

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Kerosene
Name Note low odor
MDL Number MFCD00135561
CAS 64742-47-8
UN Number UN1223
Synonym Kerosine
Health Hazard 1 Danger
Boiling Point 200° to 250°C
Health Hazard 2 GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Merck Index 14,5294
Quantity 500mL
Flash Point 76°C (169°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KEROSENE
RTECS Number OA5500000

Specifications

Chemical Name or Material Kerosene
CAS 64742-47-8
Name Note low odor
MDL Number MFCD00135561
UN Number UN1223
Merck Index 14,5294
Synonym Kerosine

Applications
Kerosene is ideal for dilution of oils and other organic fluids for determination of metal traces. It is used in paints & coatings, consumer products, printing inks and agricultural chemical applications. It is also used in food contact applications such as aluminum rolling.

Solubility
Soluble in water (0.02 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Accessories

Danger

GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.