missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kemtec™ Building Simple DC Circuits with Real Electronic Components Kit

Teach simple DC circuitry using real components and help students relate projects to real world applications.

$236.25 - $236.25

Specifications

For Use With (Application) Teach electronic circuitry using real components and help students relate projects to real life applications
Includes 5 experiments, lecture information, sample data, complete answer keys, real electronic components kit, instructor's manual
View More Specs

Alert:

Choking hazard (1) Small parts. Not for children under 3 yrs.


Includes: Red LED, multimeter, solderless breadboard, 9V battery, test lead wire, 9V battery snap, 330 Ohm resistor, 820 Ohm resistor, 560 Ohm resistor, 680 Ohm resistor, 220 Ohm (1/2 watt) resistor, 150 Ohm resistor, matches, instructor manual

Requires: Impact goggles


Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S07353 View Documents Fisher Scientific
15224
Class-pack Kit Each for $236.25
Description

Description

This basic kit introduces the use of easy-to-manipulate, reusable, solder-less breadboards, resistors, LEDs, jump wires and batteries, allowing hands-on experience with circuitry components for individual students.

  • Students learn to design circuits and read circuit diagrams
  • Students can investigate effects of resistors and LEDs in real circuits
  • Instructor's manual included with reproducible student guides covering stepwise experimental protocols, simple circuit diagrams, homework assignments and assessment materials
  • Includes separate assignments for middle and high school levels
  • Complete unit with five experiments, lecture information, sample data, and complete answer keys
  • Complete kit also includes one digital multimeter for teacher to demonstrate how to measure voltage, current and resistance so that data may be collected for problem
  • Available as single kit (1 to 4 students) or class-pack kit (24 students)
  • Grade 7 to 10

Specifications

Specifications

Teach electronic circuitry using real components and help students relate projects to real life applications
5 experiments, lecture information, sample data, complete answer keys, real electronic components kit, instructor's manual