Please login to your online account to display your discounted pricing
Kemtec™ Basic Microchemistry Kits
Microchemistry kits include all laboratory supplies required to perform chemistry experiments on microscale.
$62.10 - $62.10
Specifications
For Use With (Application) Mole Ratios, Catalysts, Rates of Reaction, Equilibrium, Titration
Includes Five experiments
View More Specs
 Alert
Choking hazard (1) Small parts. Not for children under 3 yrs.
Products
Catalog Number Mfr. No. Product Type Price Quantity    
S63132
fisher science education
4446
Basic Kit for 24 students Each for $62.10
Description & Specifications
Specifications
For Use With (Application) Mole Ratios, Catalysts, Rates of Reaction, Equilibrium, Titration
Includes Five experiments
Microchemistry sets provide an economical solution for increasing lab hour requirements with inexpensive, reusable supplies, reduced chemical costs, easy preparation and quick clean.
  • Supply of small amounts of standard laboratory chemicals is required
  • Available as single set (for 2 students) and class set (for 24 students)
  • Each class set includes supplies for twelve pairs of students, one instructor's manual and twelve student manuals
  • Student manuals feature stepwise protocols, clear instructional diagrams and questions for assessment
  • Instructor's manuals are complete with answer keys
  • Grades 9-12
Basic Microchemistry Set: Single set (Five experiments)
  • Mole Ratios
  • Catalysts
  • Rates of Reaction
  • Equilibrium
  • Titrations