missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

KD Scientific Legacy KDS 270 Continuous Cycle Pump

$9,622.85 - $9,677.83

Specifications

Flow Rate Accuracy ±<1%
Display Type Backlit LCD
Applications Electrospinning, microfluids, electrospray, mass spec, continuous flow, organic synthesis, drug infusion, Maldi-TOF, emulsifier
Weight (English) 9.5 lb.
Weight (Metric) 4 kg
View More Specs

Products 2
Catalog Number Mfr. No. Certifications/Compliance Electrical Requirements Voltage Price Quantity  
Catalog Number Mfr. No. Certifications/Compliance Electrical Requirements Voltage Price Quantity  
14-931-28 View Documents KD Scientific
780270
Not CE marked 110 VAC, 50/60 Hz 110 V Each for $9,622.85
 
14-931-30 View Documents KD Scientific
789270
CE 220 VAC 50/60 Hz 220 V Each for $9,677.83
 
Specifications

Specifications

±<1%
Electrospinning, microfluids, electrospray, mass spec, continuous flow, organic synthesis, drug infusion, Maldi-TOF, emulsifier
4 kg
Backlit LCD
9.5 lb.