Please login to your online account to display your discounted pricing

KC7F2, Tocris™

HIF-1α inhibitor; down-regulates HIF-1α protein synthesis

$116.01 - $488.83

Chemical Identifiers

Chemical Name or Material KC7F2
Assay Percent Range >98%
Molecular Formula C16H16Cl4N2O4S4
Formula Weight 570.38
Synonym N,N'-(Dithiodi-2,1-ethanediyl)bis[2,5-dichlorobenzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

432410

 
r & d systems
RDS432410
10mg Each for $116.01

432450

 
r & d systems
RDS432450
50mg Each for $488.83
Description & Specifications

Specifications

Formula Weight 570.38
Quantity 10mg
Chemical Name or Material KC7F2
Assay Percent Range >98%
Molecular Formula C16H16Cl4N2O4S4
CAS 927822-86-4
Synonym N,N'-(Dithiodi-2,1-ethanediyl)bis[2,5-dichlorobenzenesulfonamide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C