KB-R7943 mesylate, Tocris
Na+/Ca2+ exchange inhibitor (reverse mode)
Manufacturer:  R & D Systems RDS124410
 View more versions of this product
Catalog No. 124410
Specifications Description & Specifications
Specifications
KB-R7943 mesylate
182004-65-5
C16H17N3O3S·CH3SO3H
2-[2-[4-(4-Nitrobenzyloxy)phenyl]ethyl]isothiourea mesylate
10mg
>99%
427.49
Chemical Identifiers
>99%
427.49
KB-R7943 mesylate
C16H17N3O3S·CH3SO3H
2-[2-[4-(4-Nitrobenzyloxy)phenyl]ethyl]isothiourea mesylate
Safety and Handling
Recommended Storage : Store at Room Temperature