missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kasugamycin, 50mg/mL in distilled water, sterile-filtered, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J67325AC
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Kasugamycin | |
50mg/mL in distilled water, sterile-filtered | |
19408-46-9 | |
MFCD00058459 | |
ZDRBJJNXJOSCLR-YZKQBBCCSA-N | |
2-amino-2-[(2R,3S,5S,6R)-5-amino-2-methyl-6-[(2R,3S,5S,6S)-2,3,4,5,6-pentahydroxycyclohexyl]oxyoxan-3-yl]iminoacetic acid;hydron;chloride | |
66765861 |
Kasugamycin | |
Liquid | |
C14H26ClN3O9 | |
Odorless | |
[H+].CC1C(CC(C(O1)OC2C(C(C(C(C2O)O)O)O)O)N)N=C(C(=O)O)N.[Cl-] | |
415.824 | |
25 mL |
Chemical Identifiers
19408-46-9 | |
415.824 | |
ZDRBJJNXJOSCLR-YZKQBBCCSA-N | |
2-amino-2-[(2R,3S,5S,6R)-5-amino-2-methyl-6-[(2R,3S,5S,6S)-2,3,4,5,6-pentahydroxycyclohexyl]oxyoxan-3-yl]iminoacetic acid;hydron;chloride |
C14H26ClN3O9 | |
MFCD00058459 | |
66765861 | |
[H+].CC1C(CC(C(O1)OC2C(C(C(C(C2O)O)O)O)O)N)N=C(C(=O)O)N.[Cl-] |
Safety and Handling
RTECSNumber : NM7521800
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C