Please login to your online account to display your discounted pricing

Alfa Aesar™ Kasugamycin, 50mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Kasugamycin
CAS 19408-46-9
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00058459
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67325AC

  SDS
alfa aesar™
J67325AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67325AD

  SDS
alfa aesar™
J67325AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Kasugamycin
Physical Form Liquid
MDL Number MFCD00058459
CAS 19408-46-9
Name Note 50mg/mL in distilled water, sterile-filtered
Odor Odorless
Storage Note 1 -30° to -10°C
Quantity 25mL
RTECS Number NM7521800
Safety and Handling

-30° to -10°C