Please login to your online account to display your discounted pricing
Dynalon™ Kartell Weighing Bottles with Lids
Reduce the tare weight correction with these lightweight, autoclavable, reusable translucent bottles.
$79.10 - $79.10
Specifications
Closure Material Polypropylene
Color Translucent
View More Specs
Includes:  Push-on lids.
Products
Catalog Number Mfr. No. Capacity Height (English) Weight (English) Price Quantity    
S01990
dynalon™
261154-00
0.67 oz., 20mL 1.61 in. 0.01 lb. Pack of 10 Each for $79.10
Description & Specifications
Specifications
Closure Material Polypropylene
Color Translucent
  • Air-tight
  • No internal shoulder eliminates need for spatulas
  • For liquids or solids
  • Highly resistant to acid attack
  • Tops can be squeezed to assist pouring
  • Very economical compared to glass