missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Karl Fischer Solvent, solvent for volumetric two-component titration

Manufacturer:  Alfa Aesar™ 47281K2

 View more versions of this product

Catalog No. AA47281K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Karl Fischer Solvent
65°C
1L
UN3286
0.910g/mL
Colorless to Green
solvent for volumetric two-component titration
Liquid
Documents