missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Karl Fischer Solvent KT, solvent for volumetric one- and two-component titration, for aldehydes and ketones

Manufacturer:  Alfa Aesar™ 47219Y0

 View more versions of this product

Catalog No. AA47219Y0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Karl Fischer Solvent KT
120°C
6 x 1L
UN2924
0.940g/mL
Colorless to Yellow
solvent for volumetric one- and two-component titration, for aldehydes and ketones
Liquid
Documents