Please login to your online account to display your discounted pricing

Alfa Aesar™ Karl Fischer Solvent CS, solvent for volumetric titration, for oils

Chemical Identifiers

Chemical Name or Material Karl Fischer Solvent CS
Name Note solvent for volumetric titration, for oils
UN Number UN3286
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47217K2

 
alfa aesar™
47217K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47217Y0

 
alfa aesar™
47217Y0
6 x 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material Karl Fischer Solvent CS
TSCA TSCA
Name Note solvent for volumetric titration, for oils
Physical Form Liquid
UN Number UN3286
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H311-H331-H370-H351-H373-H314-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
Density 1.040g/mL
Boiling Point 59°C
Color Colorless to pale yellow
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H331-H370-H351-H373-H314-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.