Please login to your online account to display your discounted pricing

Karl Fischer Reagent (Stabilized), Fisher Chemical

$1,893.62 - $1,893.62

Chemical Identifiers

Chemical Name or Material Karl Fischer Reagent
CAS 109-86-4
CAS 110-86-1
CAS 7446-09-5
CAS 7553-56-2
Physical Form Liquid
MDL Number MFCD00081734
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SK3-4

  Accessories SDS
fisher scientific company
SK34
Amber Glass 4L Each for $1,893.62
Case of 4 Each for $7,574.48
Description & Specifications

Specifications

Boiling Point 124°C
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Treatment(s) Stabilized
Physical Form Liquid
Density 0.93g/cm³
Color Dark Brown
Chemical Name or Material Karl Fischer Reagent
CAS 7553-56-2
MDL Number MFCD00081734
CAS 109-86-4
ChemAlert Storage Symbol Red
Equivalence 6mg/mL min.
CAS 110-86-1
Packaging Amber Glass
CAS 7446-09-5

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling