Please login to your online account to display your discounted pricing

Alfa Aesar™ Karl Fischer Methanol, solvent/working medium for volumetric one-component titration

Chemical Identifiers

Chemical Name or Material Karl Fischer Methanol
Name Note solvent/working medium for volumetric one-component titration
UN Number UN1230
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47165K2

 
alfa aesar™
47165-K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47165Y0

 
alfa aesar™
47165-Y0
6 x 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 0.910g/mL
Boiling Point 65°C
Color Colorless to pale green
Quantity 1L
Chemical Name or Material Karl Fischer Methanol
Name Note solvent/working medium for volumetric one-component titration
Physical Form Liquid
UN Number UN1230
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.