missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Karl Fischer Coulometric AG-Oil, anolyte solution, for cells with or without diaphragm

$118.53 - $118.53

Chemical Identifiers

UN Number UN3286
Chemical Name or Material Karl Fischer Coulometric AG-Oil, anolyte solution, for cells with or without diaphragm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA47195AP SDS Alfa Aesar™
47195AP
500mL Each for $118.53
AA47195BA SDS Alfa Aesar™
47195BA
6 x 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

UN3286
Karl Fischer Coulometric AG-Oil, anolyte solution, for cells with or without diaphragm

Specifications

1.040g/mL
Colorless to pale yellow
Liquid
Karl Fischer Coulometric AG-Oil, anolyte solution, for cells with or without diaphragm
59°C
500mL
UN3286

Safety and Handling

GHS H Statement
H225-H311-H331-H370-H351-H373-H314-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.