Please login to your online account to display your discounted pricing

Alfa Aesar™ Karl Fischer Coulometric AG, anolyte solution, for cells with or without diaphragm

$120.09 - $623.80

Chemical Identifiers

Chemical Name or Material Karl Fischer Coulometric AG
Name Note anolyte solution, for cells with or without diaphragm
UN Number UN3286
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47106AP

  SDS
alfa aesar™
47106AP
500mL Each for $120.09

AA47106BA

  SDS
alfa aesar™
47106BA
6 x 500mL Pack for $623.80
Description & Specifications

Specifications

Quantity 500mL
Density 0.930g/mL
Boiling Point 63°C
Health Hazard 2 GHS H Statement
H225-H301-H311-H331-H370-H314
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3286
Color Colorless to pale yellow
Chemical Name or Material Karl Fischer Coulometric AG
Physical Form Liquid
Health Hazard 1 Danger
Name Note anolyte solution, for cells with or without diaphragm
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H311-H331-H370-H314
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.