Kappler™ ProVent™ 10,000 Limited-Use Lab Coats
Comfortable fit in a tailored style
Manufacturer:  Kappler™ MS235BUMD
 View more versions of this product
Catalog No. 17-988-492B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Patented microporous film composite fabric offers a wide range of barrier protection against blood, body fluids and viral threats.
  • High moisture vapor transmission rate provides cool, comfort
  • Ideal for biohazard cleanup
  • Six snaps for front closure
  • Serged-seam stitching, breast pocket, two hip pockets
  • Knit collar and cuffs
Compliance
Passes ASTM™ 1670 blood penetration and ASTM 1671 viral penetration tests
Specifications
Specifications
Yes
ASTM 1670, ASTM 1671
Blue
3
Serged
8.2 kg
Non-sterile
Snaps
ProVent™ fabric
Medium
18 lb.