missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kappler™ ProVent™ 10,000 Limited-Use Lab Coats
Comfortable fit in a tailored style
Manufacturer: Kappler™ MS235WHMD
Description
Patented microporous film composite fabric offers a wide range of barrier protection against blood, body fluids and viral threats.
- High moisture vapor transmission rate provides cool, comfort
- Ideal for biohazard cleanup
- Six snaps for front closure
- Serged-seam stitching, breast pocket, two hip pockets
- Knit collar and cuffs
Compliance
Passes ASTM™ 1670 blood penetration and ASTM 1671 viral penetration tests
Specifications
Yes | |
ASTM 1670, ASTM 1671 | |
White | |
ProVent™ fabric | |
Medium | |
18 lb. |
Non-sterile | |
Snaps | |
Knit | |
3 | |
Serged | |
8.2 kg |