missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

kappa-Carrageenan, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC431591000

 View more versions of this product

Catalog No. AC431591000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

White to almost white to yellow to brownish
100g
Authentic
κ-Carrageenan
11114-20-8
Glass bottle
Powder
10mPa&middnot;s Min. (0.3% water, 25°C)
Irish moss
MFCD00081480
40% max.
12% max. (1 g, 105°C)

Chemical Identifiers

11114-20-8
MFCD00081480
Irish moss
κ-Carrageenan

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning