missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kaolin, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 211740025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Kaolin | |
Beige | |
Powder | |
Al2H4O9Si2 | |
14,5282 | |
Solubility in water: insoluble. Other solubilities: insoluble in cold acids and alkali hydroxides | |
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O | |
258.16 | |
258.2 | |
1.5% max. | |
Plastic Bottle |
1750.0°C | |
1332-58-7 | |
2.5 kg | |
MFCD00062311 | |
kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss | |
NLYAJNPCOHFWQQ-UHFFFAOYSA-N | |
oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate | |
56841936 | |
Pure | |
10.8% to 11.8% |
Chemical Identifiers
1332-58-7 | |
258.16 | |
NLYAJNPCOHFWQQ-UHFFFAOYSA-N | |
56841936 | |
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O |
Al2H4O9Si2 | |
MFCD00062311 | |
kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss | |
oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate |
RUO – Research Use Only