missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kanamycin disulfate, 700μg/mg, Thermo Scientific Chemicals
$170.44 - $170.44
Chemical Identifiers
CAS | 64013-70-3 |
---|---|
Molecular Formula | C18H40N4O19S2 |
Molecular Weight (g/mol) | 680.647 |
MDL Number | MFCD00143916 |
InChI Key | OGTKIXVMLDAMNU-KNQICTBBSA-N |
Synonym | Kanamycin A; Kanamycin acid sulfate |
PubChem CID | 24721551 |
IUPAC Name | (2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid |
SMILES | C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6685606
|
Thermo Scientific Chemicals
J6685606 |
5 g |
Each for $170.44
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64013-70-3 | |
680.647 | |
OGTKIXVMLDAMNU-KNQICTBBSA-N | |
24721551 | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O |
C18H40N4O19S2 | |
MFCD00143916 | |
Kanamycin A; Kanamycin acid sulfate | |
(2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid |
Specifications
Kanamycin Disulfate | |
64013-70-3 | |
C18H40N4O19S2 | |
Kanamycin A; Kanamycin acid sulfate | |
OGTKIXVMLDAMNU-KNQICTBBSA-N | |
(2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid | |
24721551 | |
Colorless | |
Light and moisture sensitive |
Powder | |
>260°C | |
MFCD00143916 | |
Soluble in aqueous solutions | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O | |
680.647 | |
680.65 | |
5 g | |
Kanamycin disulfate, 700μg/mg |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H360
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only