Please login to your online account to display your discounted pricing

Alfa Aesar™ Kanamycin, 50mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Kanamycin
CAS 25389-94-0
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00070289
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67354AD

  SDS
alfa aesar™
J67354AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67354AE

  SDS
alfa aesar™
J67354AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Kanamycin
MDL Number MFCD00070289
CAS 25389-94-0
Health Hazard 3 GHS P Statement
P285-P261-P280-P342+P311-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Physical Form Liquid
Storage Note 1 -30° to -10°C
Health Hazard 2 GHS H Statement
H334-H360-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage fertility or the unborn child.
May cause an allergic skin reaction.
Name Note 50mg/mL in distilled water, sterile-filtered
Quantity 50mL

Applications
It has been used in tissue culture media. It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium. It can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100mg/mL. Kanamycin can be used as a selection agent in several types of isolation media.

Solubility
Fully miscible in water. Insoluble in acetone, chloroform, ether, ethyl acetate, and alcohol.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

Danger

GHS P Statement
P285-P261-P280-P342+P311-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H360-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage fertility or the unborn child.
May cause an allergic skin reaction.

-30° to -10°C