Please login to your online account to display your discounted pricing

Kainic acid, Tocris


Kainic acid, Tocris

Quantity 50mg

Manufacturer: r & d systems  022250

 View more versions of this product

Catalog No. 022250

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Physical Form Solid
Formula Weight 213.23
CAS 487-79-6
Molecular Formula C10H15NO4
Assay Percent Range >98%
Synonym (2S, 3S, 4S)-Carboxy-4-(1-methylethenyl)-3-pyrrolidineacetic acid
Recommended Storage Store at Room Temperature
Chemical Name or Material Kainic acid

Specifications

Physical Form Solid
Accessories

Store at Room Temperature