Please login to your online account to display your discounted pricing

Kainic acid, Tocris

Kainic acid, Tocris

$834.54 - $834.54

Chemical Identifiers

Physical Form Solid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

022250

 
r & d systems
022250
50mg Each for $834.54
Description & Specifications

Specifications

Quantity 50mg
Physical Form Solid
Formula Weight 213.23
Chemical Name or Material Kainic acid
Assay Percent Range >98%
Molecular Formula C10H15NO4
CAS 487-79-6
Synonym (2S, 3S, 4S)-Carboxy-4-(1-methylethenyl)-3-pyrrolidineacetic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature