Please login to your online account to display your discounted pricing

Kaempferol, 99.00%, MP Biomedicals™


Quantity 10mg

Manufacturer: mp biomedicals  15514310

 View more versions of this product

Catalog No. ICN15514310

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 286.2
Color Yellow
Quantity 10mg
Chemical Name or Material Kaempferol
Molecular Formula C15H10O6
CAS 520-18-3
Synonym Nimbecetin

Specifications

Chemical Name or Material Kaempferol
CAS 520-18-3
Physical Form Powder
Molecular Formula C15H10O6
Formula Weight 286.2
Synonym Nimbecetin