Please login to your online account to display your discounted pricing

Kaempferol, 99.00%, MP Biomedicals™

$70.25 - $402.05

Chemical Identifiers

Chemical Name or Material Kaempferol
CAS 520-18-3
Physical Form Powder
Molecular Formula C15H10O6
Formula Weight 286.2
Synonym Nimbecetin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15514310

 
mp biomedicals
15514310
10mg Each for $70.25

ICN15514380

 
mp biomedicals
15514380
100mg Each for $402.05
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 286.2
Color Yellow
Quantity 10mg
Chemical Name or Material Kaempferol
Molecular Formula C15H10O6
CAS 520-18-3
Synonym Nimbecetin