missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

K 579, Tocris

Dipeptidyl peptidase IV (DPP-IV) inhibitor

$290.95 - $1,205.80

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C17H24N6O
Formula Weight 328.41
Synonym (2S)-1-[[[4-Methyl-1-(2-pyrimidinyl)-4-piperidinyl]amino]acetyl]-2-pyrrolidinecarbonitrile
Chemical Name or Material K 579
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
279010 R & D Systems
279010
10mg Each for $290.95
279050 R & D Systems
279050
50mg Each for $1,205.80
Specifications Description & Specifications

Chemical Identifiers

>99%
328.41
K 579
C17H24N6O
(2S)-1-[[[4-Methyl-1-(2-pyrimidinyl)-4-piperidinyl]amino]acetyl]-2-pyrrolidinecarbonitrile

Specifications

10mg
>99%
328.41
K 579
440100-64-1
C17H24N6O
(2S)-1-[[[4-Methyl-1-(2-pyrimidinyl)-4-piperidinyl]amino]acetyl]-2-pyrrolidinecarbonitrile

Safety and Handling

Recommended Storage : Desiccate at +4°C