Please login to your online account to display your discounted pricing

K 114, Tocris

Amyloid fibril-specific fluorescent dye

$585.69 - $585.69

Chemical Identifiers

Chemical Name or Material K 114
Assay Percent Range >99%
Molecular Formula C22H17BrO2
Formula Weight 393.27
Synonym 4,4'-[(2-Bromo-1,4-phenylene)di-(1E)-2,1-ethenediyl]bisphenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

314450

 
r & d systems
RDS314450
50mg Each for $585.69
Description & Specifications

Specifications

Formula Weight 393.27
Quantity 50mg
Chemical Name or Material K 114
Molecular Formula C22H17BrO2
CAS 872201-12-2
Synonym 4,4'-[(2-Bromo-1,4-phenylene)di-(1E)-2,1-ethenediyl]bisphenol
Recommended Storage Store at +4°C
Assay Percent Range >99%
Safety and Handling

Store at +4°C