Please login to your online account to display your discounted pricing

JX 401, Tocris

Potent, reversible p38α inhibitor

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material JX 401
Assay Percent Range >98%
Molecular Formula C21H25NO2S
Formula Weight 355.49
Synonym 1-[2-Methoxy-4-(methylthio)benzoyl]-4-(phenylmethyl)piperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

265710

 
r & d systems
RDS265710
10mg Each for $239.63

265750

 
r & d systems
RDS265750
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 355.49
Quantity 10mg
Synonym 1-[2-Methoxy-4-(methylthio)benzoyl]-4-(phenylmethyl)piperidine
Chemical Name or Material JX 401
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C21H25NO2S
CAS 349087-34-9
Safety and Handling

Store at Room Temperature