Please login to your online account to display your discounted pricing

JW 55, Tocris™

Tankyrase inhibitor; inhibits canonical Wnt signaling

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material JW 55
Assay Percent Range >98%
Molecular Formula C25H26N2O5
Formula Weight 434.48
Synonym N-[4-[[[[Tetrahydro-4-(4-methoxyphenyl)-2H-pyran-4-yl]methyl]amino]carbonyl]phenyl]-2-furancarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

451410

 
r & d systems
RDS451410
10mg Each for $239.63

451450

 
r & d systems
RDS451450
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 434.48
Quantity 10mg
Assay Percent Range >98%
CAS 664993-53-7
Molecular Formula C25H26N2O5
Synonym N-[4-[[[[Tetrahydro-4-(4-methoxyphenyl)-2H-pyran-4-yl]methyl]amino]carbonyl]phenyl]-2-furancarboxamide
Chemical Name or Material JW 55
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C