Please login to your online account to display your discounted pricing

JW 480, Tocris™

Potent and selective inhibitor of serine hydrolase KIAA1363 (AADACL1)

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material JW 480
Assay Percent Range >98%
Molecular Formula C22H23NO2
Formula Weight 333.42
Synonym 2-Isopropylphenyl(2-(naphthalen-2-yl)ethyl)carbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

432710

 
r & d systems
RDS432710
10mg Each for $139.08

432750

 
r & d systems
RDS432750
50mg Each for $585.69
Description & Specifications

Specifications

Chemical Name or Material JW 480
Quantity 10mg
CAS 1354359-53-7
Recommended Storage Store at -20°C
Assay Percent Range >98%
Molecular Formula C22H23NO2
Formula Weight 333.42
Synonym 2-Isopropylphenyl(2-(naphthalen-2-yl)ethyl)carbamate
Safety and Handling

Store at -20°C