missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Sure-Grip™ EX Flammable Safety Cabinet

Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.

Manufacturer:  Justrite™ 893000

 View more versions of this product

Catalog No. 19-120-2333



Description

Description

Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.

  • Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
  • Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
  • Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self-latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)

  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Meet OSHA and NFPA requirements

Certifications

FM approved except 19-120-2336.

Compliance

Meet NFPA Code 30 and OSHA regulations; models with self-close doors also comply with the Uniform Fire Code.

Warranty

10-Year

Specifications

Specifications

30 gal.
14.56 in.
37cm
Manual-close
39.62 in.
100.6cm
2
Epoxy, Polyester
231 lb.
39.5 in.
100.3cm
18 in.
Steel (CR)
FM, NFPA, OSHA
45.7cm
14.56 x 39.5 x 39.62 in. (37 x 100.3 x 100.6cm)
Flammable liquid storage and protection
112cm
U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with
1
Can storage cabinet
105kg
109cm
Yellow
44 in.
43 in.
SDS
Documents