missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Standard Aerosolv™ Aerosol Can Disposal System: Cartridges and Filters
For use with Serosolv Can Disposal System
Manufacturer: Justrite™ 28223
Description
Replaxement cartridges and filters.
Specifications
EPA, 40CRF261.23(a)(6) | |
For aerosolv aerosol can disposal system | |
Carbon | |
Color-Changing Activated Carbon Cartridge |