missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Nonmetallic Oval Type I Safety Cans For Flammables

30% more storage capacity than round shape models.

$159.23 - $159.23

Specifications

Certifications/Compliance FM, NFPA, OSHA
Diameter (English) Opening 1.45 in.
Diameter (Metric) Opening 3.6cm
Material PE (Polyethylene)
Product Type Safety Can
View More Specs

Products 1
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
S04768MF View Documents Justrite™
14160EMD
Red Each for $159.23
 
Description

Description

  • Constructed of high-density polyethylene
  • Trigger mechanism enables pouring of small quantities of liquid
  • Leak-tight, self-closing cap prevents spillage and automatically vents to guard against explosion
  • Dual-density flame arrester protects against external heat sources
  • Current carrying carbon insert imbedded into can for proper grounding
  • Nonmetallic, stainless-steel hardware

Specifications

Specifications

FM, NFPA, OSHA
3.6cm
Safety Can
Dual density flame arrester, self closing cap, Stainless Steel hardware
Oval
1.45 in.
PE (Polyethylene)
For flammables
Dent, puncture and chip resistant