missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Nonmetallic Oval Type I Safety Cans For Flammables

30% more storage capacity than round shape models.

Manufacturer:  Justrite™ 14160EMD

 View more versions of this product

Catalog No. S04768MF



Description

Description

  • Constructed of high-density polyethylene
  • Trigger mechanism enables pouring of small quantities of liquid
  • Leak-tight, self-closing cap prevents spillage and automatically vents to guard against explosion
  • Dual-density flame arrester protects against external heat sources
  • Current carrying carbon insert imbedded into can for proper grounding
  • Nonmetallic, stainless-steel hardware

Specifications

Specifications

FM, NFPA, OSHA
1.45 in.
For flammables
32.3cm
7.6 in.
Safety Can
4.6 in.
1 gal.
Red
Dent, puncture and chip resistant
3.6cm
12.75 in.
Dual density flame arrester, self closing cap, Stainless Steel hardware
19.3cm
Oval
11.6cm
4L
PE (Polyethylene)
Documents