Justrite™ Sure-Grip™ EX Slimline Flammable Safety Cabinet
Add storage in tight spaces by adding a Justrite Sure-Grip™ EX Slimline Flammable Safety Cabinet to your lab.
Manufacturer:  Justrite™ 892200*
Catalog No. 29-635-2459
Description
Description
Double-wall, air-insulated, welded steel construction offers fire resistance in a compact design.
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • 2 in. (5cm) leakproof sump controls spills
  • With manual or self-closing door
Order Info
Capacity: 22 gal. (83L). Dimensions: 18L × 23.25W × 65 in.H (46 × 59 × 165cm).
Certifications
FM approved.
Compliance
Meet OSHA and NFPA requirements. Self-closing model complies with Uniform Fire Code.
Warranty
10-Year