missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Sure-Grip™ EX Slimline Flammable Safety Cabinet

Add storage in tight spaces by adding a Justrite Sure-Grip™ EX Slimline Flammable Safety Cabinet to your lab.

Supplier:  Justrite™ 892220

 View more versions of this product

Catalog No. 19160387


Description

Description

Double-wall, air-insulated, welded steel construction offers fire resistance in a compact design.

  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • 2 in. (5cm) leakproof sump controls spills
  • With manual or self-closing door

Order Info

Capacity: 22 gal. (83L). Dimensions: 18L x 23.25W x 65 in.H (46 x 59 x 165cm).

Certifications

FM approved.

Compliance

Meet OSHA and NFPA requirements. Self-closing model complies with Uniform Fire Code.

Warranty

10-Year

Specifications

Specifications

50.8 cm
65 in.
Steel (CR)
90 kg
14.56 in.
45.7 cm
22 gal.
20 in.
153.7 cm
Flammables
59.05 cm
165.1 cm
1
37 cm
Epoxy, Polyester
U-Loc™ handle with cylinder lock and key set, 3 Galvanized Steel shelves and 2 in. leakproof sump, dual vents, adjustable leveling feet, a grounding connector and large trilingual warning label
18 in.
14.56 x 20 x 60.5 in. (37 x 50.8 x 153.7 cm)
200 lb.
3
60.5 in.
Slimline storage cabinet
23.25 in.
Yellow
Self-close
FM, NFPA, OSHA, U (NFPA)
SDS
Documents

Documents

Promotions

Promotions