missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Sure-Grip™ EX Flammable Safety Cabinet

When using a Sure-Grip EX Flammable Safety Cabinets, these two-door and bifold door cabinets protect workers, reduce fire risks, and improve productivity by allowing storage of flammable fuels and chemicals in code-compliant, sturdy 18-gauge steel structures.

Manufacturer:  Justrite™ 893300

 View more versions of this product

Catalog No. 29-635-6144



Description

Description

Features of Justrite Flammables Cabinets

  • Double-wall construction provides 1.5 in. of insulating air space to help prevent the spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents

  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Lettering on door reads: “FLAMMABLE—KEEP FIRE AWAY”
  • Accepts padlock (not included)
  • Color: yellow, gray (optional), white (19-160-376)

Certifications

FM approved.

Compliance

Meet NFPA Code 30 and comply with OSHA regulations.

Warranty

10 years

Specifications

Specifications

30 gal.
21.75 in.
55.2cm
Manual-close
30.62 in.
77.8cm
2
Epoxy, Polyester
204 lb.
39.62 in.
82.9cm
24 in.
Steel (CR)
36 in.
FM, NFPA, OSHA, U (NFPA)
610cm
21.75 x 39.62 x 30.62 in. (55.2 x 82.9 x 77.8cm)
Flammable liquid storage and protection
88.9cm
U-Loc™ handle with two keys and accept padlock, Haz-Alert™ reflective exterior labeling and Galvanized steel SpillSlope™ shelves, dual vents with flame arresters, four adjustable self leveling feet, gr
1
Can storage cabinet
93kg
91.44cm
Yellow
35 in.
Safety
Documents