Justrite™ Coalescing Carbon Filters
Use with Justrite Quick-Disconnect Safety Cans
Manufacturer:  Justrite™ 28162
 View more versions of this product
Catalog No. 19-052-373
Description
Description
  • Ideal for HPLC applications—minimize odor by capturing 99% of liquid particulates released in the form of gaseous vapor
  • Two-stage vent filter is easy to install, snapping conveniently into factory installed quick-disconnect fitting
  • Color-changing carbon cartridge (top portion) changes from purple to black as hydrocarbons are absorbed, indicating when replacement is necessary
  • Coalescing filter (bottom portion) can be reused with new carbon filters several times and replaced after 3-4 carbon filter cycles
Cannot be used with 1-gal. oval style cans. Do not transport safety can while carbon filter is attached.
Compliance
Meets EPA 40 CFR 264.173
When using HPLC carbon filter with FM approved Justrite safety can: The Justrite safety disposal cans have not been FM approved or tested in assembly with HPLC filters attached. However, the can does comply with OSHA 1910 and NFPA Code 30 requirements, both with and without the HPLC filter.
Specifications
Specifications
EPA 40 CFR 264.173
27.94cm
4 in.
HPLC
Plastic
11 in.
Coalescing Carbon Filter
10.16cm
Justrite quick disconnect cans with stainless steel fittings