Please login to your online account to display your discounted pricing

JNJ 7777120, Tocris™

Selective H4 receptor antagonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material JNJ 7777120
Assay Percent Range >98%
Molecular Formula C14H16ClN3O
Formula Weight 277.75
Synonym 1-[(5-Chloro-1H-indol-2-yl)carbonyl]-4-methylpiperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

402110

 
r & d systems
402110
10mg Each for $205.70

402150

 
r & d systems
402150
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 277.75
Quantity 10mg
Chemical Name or Material JNJ 7777120
Assay Percent Range >98%
Molecular Formula C14H16ClN3O
CAS 459168-41-3
Synonym 1-[(5-Chloro-1H-indol-2-yl)carbonyl]-4-methylpiperazine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C